siRNA Details
| Gene Symbol: Rpe65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029987.2 | NP_084263.2 | 7 | 755 | ||
| GenBank mRNA | AF410461.1 | - | 6 | 703 | ||
| AK020906.1 | - | 7 | 754 | |||
| BC090656.1 | - | 7 | 731 | |||
| BC130028.1 | - | 6 | 707 | 40140883 | ||

Rpe65
Genomic Map