siRNA Details
| Gene Symbol: Rph3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB112924.1 | - | 5 | 398 | ||
| AK139040.1 | - | 4 | 340 | |||
| BC042585.1 | - | 5 | 565 | 5364086 | ||
| BC050883.1 | - | 5 | 532 | 5698459 | ||
| D29965.1 | - | 5 | 375 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rph3a| Gene Symbol: Rph3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB112924.1 | - | 5 | 398 | ||
| AK139040.1 | - | 4 | 340 | |||
| BC042585.1 | - | 5 | 565 | 5364086 | ||
| BC050883.1 | - | 5 | 532 | 5698459 | ||
| D29965.1 | - | 5 | 375 | |||