siRNA Details
| Gene Symbol: Rsu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009105.4 | NP_033131.2 | 5 | 554 | ||
| GenBank mRNA | AK009288.1 | - | 5 | 447 | ||
| AK011857.1 | - | 5 | 447 | |||
| AK013076.1 | - | 1 | 25 | |||
| AK135491.1 | - | 6 | 537 | |||
| AK149835.1 | - | 5 | 424 | |||
| AK150011.1 | - | 5 | 422 | |||
| AK152655.1 | - | 5 | 434 | |||
| AK169076.1 | - | 5 | 472 | |||
| BC003827.1 | - | 5 | 417 | 3493122 | ||
| CX228720.1 | - | 5 | 554 | |||

Rsu1
Genomic Map