siRNA Details
| Gene Symbol: S100b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009115.3 | NP_033141.1 | 2 | 230 | ||
| GenBank mRNA | AK134625.1 | - | 2 | 157 | ||
| AK135083.1 | - | 2 | 213 | |||
| AK161945.1 | - | 2 | 206 | |||
| AW456919.1 | - | 3 | 302 | |||
| BC061178.1 | - | 2 | 213 | 6535966 | ||
| CX205737.1 | - | 2 | 230 | |||

S100b
Genomic Map