siRNA Details
Gene Symbol: S100b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009115.3 | NP_033141.1 | 3 | 289 | ||
GenBank mRNA | AK134625.1 | - | 3 | 216 | ||
AK135083.1 | - | 3 | 272 | |||
AK161945.1 | - | 3 | 265 | |||
AW456919.1 | - | 2 | 243 | |||
BC061178.1 | - | 3 | 272 | 6535966 | ||
CX205737.1 | - | 3 | 289 |
