siRNA Details
Gene Symbol: Scn9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290674.1 | NP_001277603.1 | 23, 24 | 4802 | ||
NM_001290675.1 | NP_001277604.1 | 23, 24 | 4769 | |||
GenBank mRNA | BC158048.1 | - | 23, 24 | 4460 | 9088161 | |
BC172147.1 | - | 23, 24 | 4493 | 9054817 |
