siRNA Details
| Gene Symbol: Sct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287171.1 | NP_001274100.1 | 3 | 292 | ||
| NM_011328.3 | NP_035458.1 | 3 | 292 | |||
| GenBank mRNA | AK145859.1 | - | 3 | 291 | ||
| BC048484.1 | - | 3 | 292 | 6707701 | ||
| CF616255.1 | - | 263 | ||||
| X73580.1 | - | 3 | 265 | |||

Sct
Genomic Map