siRNA Details
| Gene Symbol: Ccl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011329.3 | NP_035459.1 | 3 | 290 | ||
| GenBank mRNA | AK019040.1 | - | 3 | 290 | ||
| BC042744.1 | - | 1901 | ||||
| BC120806.1 | - | 3 | 281 | 40129729 | ||
| M17957.1 | - | 3 | 293 | |||
| M23501.1 | - | 3 | 389 | |||

Ccl1
Genomic Map