siRNA Details
| Gene Symbol: Frrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113478.1 | NP_001106950.1 | 11, 12 | 1411 | ||
| NM_009146.3 | NP_033172.2 | 11, 12 | 1373 | |||
| GenBank mRNA | AK075573.1 | - | 11, 12 | 1365 | ||
| AK144933.1 | - | 11, 12 | 1365 | |||
| AK162693.1 | - | 11, 12 | 1366 | |||
| AK167289.1 | - | 11, 12 | 1401 | |||
| BC027770.1 | - | 10, 11 | 1371 | 5344922 | ||
| D50464.1 | - | 11, 12 | 1361 | |||

Frrs1
Genomic Map