siRNA Details
| Gene Symbol: Selplg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009151.3 | NP_033177.3 | 2 | 1264 | ||
| GenBank mRNA | AK089188.1 | - | 2 | 1240 | ||
| AK155762.1 | - | 2 | 1198 | |||
| AK156933.1 | - | 2 | 1195 | |||
| AK158134.1 | - | 2 | 1196 | |||
| AK171693.1 | - | 3 | 999 | |||
| AK172077.1 | - | 2 | 1198 | |||
| BC003874.1 | - | 3 | 1136 | 3495181 | ||
| BG070861.2 | - | 1 | 585 | |||
| BG083864.2 | - | 1 | 435 | |||
| X91144.1 | - | 2 | 1053 | |||

Selplg
Genomic Map