siRNA Details
| Gene Symbol: Sema6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018744.2 | NP_061214.2 | 13 | 2017 | ||
| GenBank mRNA | AF030430.1 | - | 13 | 1421 | ||
| AK042751.1 | - | 13 | 2018 | |||
| AK078490.1 | - | 1 | 3322 | |||
| AK082711.1 | - | 13 | 2020 | |||
| BC059238.1 | - | 13 | 1979 | 6417475 | ||
| BC062979.1 | - | 13 | 1595 | 6841689 | ||

Sema6a
Genomic Map