siRNA Details
| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009164.2 | NP_033190.2 | 8 | 785 | ||
| GenBank mRNA | AK145156.1 | - | 8 | 725 | ||
| AK161215.1 | - | 7 | 709 | |||
| AK166337.1 | - | 8 | 778 | |||
| AK172060.1 | - | 8 | 782 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3bp1| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009164.2 | NP_033190.2 | 8 | 785 | ||
| GenBank mRNA | AK145156.1 | - | 8 | 725 | ||
| AK161215.1 | - | 7 | 709 | |||
| AK166337.1 | - | 8 | 778 | |||
| AK172060.1 | - | 8 | 782 | |||