siRNA Details
| Gene Symbol: Itsn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198968.2 | NP_001185897.1 | 8 | 984 | ||
| NM_001198969.2 | NP_001185898.1 | 8 | 984 | |||
| NM_011365.4 | NP_035495.2 | 8 | 984 | |||
| GenBank mRNA | AF132479.1 | - | 7 | 724 | ||
| AF132480.1 | - | 7 | 724 | |||
| AK029325.1 | - | 8 | 931 | |||
| AK034367.1 | - | 8 | 985 | |||
| AK042449.1 | - | 8 | 954 | |||
| AK122480.1 | - | 3 | 371 | |||
| AK163807.1 | - | 8 | 958 | |||
| BC138262.1 | - | 8 | 961 | 8861281 | ||
| BC171950.1 | - | 8 | 961 | 9054236 | ||

Itsn2
Genomic Map