siRNA Details
| Gene Symbol: Shfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009169.2 | NP_033195.1 | 1 | 133 | ||
| GenBank mRNA | AK135462.1 | - | 85 | |||
| AK141911.1 | - | 1 | 139 | |||
| AK160432.1 | - | 1 | 126 | |||
| AK161690.1 | - | 1 | 106 | |||
| AK167914.1 | - | 1 | 83 | |||
| BC058117.1 | - | 1 | 99 | 6412144 | ||

Shfm1
Genomic Map