siRNA Details
| Gene Symbol: Shfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009169.2 | NP_033195.1 | 1 | 165 | ||
| GenBank mRNA | AK135462.1 | - | 117 | |||
| AK141911.1 | - | 1 | 171 | |||
| AK160432.1 | - | 1 | 158 | |||
| AK161690.1 | - | 1 | 138 | |||
| AK167914.1 | - | 1 | 115 | |||
| BC023490.1 | - | 1 | 122 | 5251089 | ||
| BC058117.1 | - | 1 | 131 | 6412144 | ||
| U41626.1 | - | 1 | 107 | |||
| X83589.1 | - | 1 | 141 | |||

Shfm1
Genomic Map