siRNA Details
| Gene Symbol: Six6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011384.4 | NP_035514.1 | 1 | 243 | ||
| GenBank mRNA | AF050130.2 | - | 1 | 278 | ||
| AF135267.1 | - | 1 | 294 | |||
| AJ011787.1 | - | 1 | 132 | |||
| AK017544.1 | - | 1 | 307 | |||
| AK029309.1 | - | 1 | 244 | |||
| BC138838.1 | - | 1 | 216 | 8861860 | ||
| BC138839.1 | - | 1 | 216 | 8861861 | ||

Six6
Genomic Map