siRNA Details
| Gene Symbol: Six6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011384.4 | NP_035514.1 | 1 | 315 | ||
| GenBank mRNA | AF050130.2 | - | 1 | 350 | ||
| AF135267.1 | - | 1 | 366 | |||
| AJ011787.1 | - | 1 | 204 | |||
| AK017544.1 | - | 1 | 379 | |||
| AK029309.1 | - | 1 | 316 | |||
| BC138838.1 | - | 1 | 288 | 8861860 | ||
| BC138839.1 | - | 1 | 288 | 8861861 | ||

Six6
Genomic Map