siRNA Details
| Gene Symbol: Skil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039090.2 | NP_001034179.1 | 2 | 945 | ||
| NM_011386.3 | NP_035516.2 | 2 | 945 | |||
| GenBank mRNA | AB221599.1 | - | 1 | 222 | ||
| AK087797.1 | - | 2 | 894 | |||
| AK154471.1 | - | 2 | 892 | |||
| AK162952.1 | - | 2 | 944 | |||
| AK163526.1 | - | 2 | 872 | |||
| AK171402.1 | - | 2 | 894 | |||
| BC049934.1 | - | 1 | 371 | 5387475 | ||
| U10530.1 | - | 1 | 716 | |||
| U10532.1 | - | 1 | 716 | |||
| U14655.1 | - | 1 | 278 | |||

Skil
Genomic Map