siRNA Details
| Gene Symbol: Slc12a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079690.1 | NP_001073158.1 | 2 | 283 | ||
| NM_183354.2 | NP_899197.2 | 2 | 283 | |||
| GenBank mRNA | AK052750.1 | - | 2 | 304 | ||
| AK085455.1 | - | 1 | 1380 | |||
| AK128928.1 | - | 2 | 280 | |||
| BC016888.1 | - | 1 | 261 | 4236020 | ||
| U20973.1 | - | 2 | 281 | |||
| U20974.1 | - | 2 | 281 | |||
| U20975.1 | - | 2 | 281 | |||
| U61381.1 | - | 1 | 252 | |||
| U94518.1 | - | 1 | 230 | |||

Slc12a1
Genomic Map