siRNA Details
| Gene Symbol: Slc22a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011396.3 | NP_035526.1 | 6 | 1186 | ||
| GenBank mRNA | AB015800.1 | - | 6 | 1081 | ||
| AF110417.1 | - | 6 | 1134 | |||
| AF111425.1 | - | 6 | 1121 | |||
| AK140640.1 | - | 6 | 1182 | |||
| AK167451.1 | - | 6 | 1162 | |||
| BC031118.1 | - | 6 | 1143 | 4981427 | ||

Slc22a5
Genomic Map