siRNA Details
| Gene Symbol: Slc22a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009203.3 | NP_033229.3 | 9 | 1762 | ||
| GenBank mRNA | AB005451.1 | - | 9 | 1543 | ||
| AI314749.1 | - | 2 | 465 | |||
| AK142580.1 | - | 8 | 911 | |||
| BC035927.1 | - | 1698 | 4242354 | |||

Slc22a12
Genomic Map