siRNA Details
| Gene Symbol: Slc31a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175090.4 | NP_780299.2 | 5 | 713 | ||
| GenBank mRNA | AK046835.1 | - | 5 | 666 | ||
| AK151337.1 | - | 5 | 695 | |||
| AK170285.1 | - | 5 | 666 | |||
| BC034674.1 | - | 5 | 651 | 3968084 | ||
| BC046470.1 | - | 3 | 387 | |||
| BC058227.1 | - | 5 | 653 | 6309323 | ||
| BC065147.1 | - | 3 | 1123 | |||

Slc31a1
Genomic Map