siRNA Details
| Gene Symbol: Slfn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137559.1 | - | 6 | 1273 | ||
| AK137689.1 | - | 5 | 1194 | |||
| BC044865.1 | - | 6 | 1282 | 4459564 | ||
| BC150770.1 | - | 3 | 889 | 9087681 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slfn4| Gene Symbol: Slfn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137559.1 | - | 6 | 1273 | ||
| AK137689.1 | - | 5 | 1194 | |||
| BC044865.1 | - | 6 | 1282 | 4459564 | ||
| BC150770.1 | - | 3 | 889 | 9087681 | ||