siRNA Details
| Gene Symbol: Snai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011415.2 | NP_035545.1 | 2 | 570 | ||
| GenBank mRNA | AK133968.1 | - | 2 | 578 | ||
| BC062164.1 | - | 2 | 570 | 30242610 | ||
| U79550.1 | - | 2 | 498 | |||
| U97059.1 | - | 2 | 455 | |||

Snai2
Genomic Map