siRNA Details
| Gene Symbol: Smr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252679.1 | NP_001239608.1 | 5 | 601 | ||
| NM_001252680.1 | NP_001239609.1 | 4 | 531 | |||
| NM_001252681.1 | NP_001239610.1 | 3 | 464 | |||
| NM_021289.2 | NP_067264.2 | 3 | 464 | |||
| GenBank mRNA | BC111829.1 | - | 4 | 494 | 40055208 | |
| U82376.1 | - | 4 | 531 | |||
| U82377.1 | - | 3 | 464 | |||
| U82378.1 | - | 3 | 464 | |||
| U82379.1 | - | 5 | 601 | |||

Smr2
Genomic Map