siRNA Details
| Gene Symbol: Sntb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009229.4 | NP_033255.1 | 5 | 1187 | ||
| GenBank mRNA | AK040423.1 | - | 5 | 1188 | ||
| AK080059.1 | - | 5 | 1184 | |||
| AK131687.1 | - | 5 | 1212 | |||
| AK165009.1 | - | 5 | 1189 | |||
| AK165900.1 | - | 5 | 1164 | |||
| BC030352.1 | - | 3 | 544 | |||
| BC038704.1 | - | 3 | 475 | |||
| BC138850.1 | - | 5 | 1186 | 8861872 | ||
| BC145442.1 | - | 5 | 1186 | 9054000 | ||
| U00678.1 | - | 5 | 1186 | |||

Sntb2
Genomic Map