siRNA Details
| Gene Symbol: Son | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019973.2 | NP_064357.2 | 2 | 246 | ||
| NM_178880.4 | NP_849211.3 | 2 | 246 | |||
| GenBank mRNA | AB546195.1 | - | 2 | 181 | ||
| AF193607.1 | - | 2 | 190 | |||
| AK008256.1 | - | 2 | 187 | |||
| AK008478.1 | - | 2 | 187 | |||
| AK019081.1 | - | 2 | 194 | |||
| AK019312.1 | - | 2 | 218 | |||
| AK161714.1 | - | 1 | 731 | |||
| BC046419.1 | - | 2 | 185 | |||
| BC051200.1 | - | 2 | 191 | |||
| BC055728.1 | - | 2 | 172 | |||
| BC061113.1 | - | 2 | 202 | |||
| BC145121.1 | - | 2 | 186 | |||

Son
Genomic Map