siRNA Details
| Gene Symbol: Sp100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013673.3 | NP_038701.2 | 4, 5 | 483 | ||
| GenBank mRNA | AK042095.1 | - | 4, 5 | 519 | ||
| AK080004.1 | - | 4, 5 | 520 | |||
| AK083304.1 | - | 4, 5 | 484 | |||
| AK146054.1 | - | 4, 5 | 520 | |||
| AK152348.1 | - | 4, 5 | 520 | |||
| AK153487.1 | - | 4, 5 | 520 | |||
| BC069183.1 | - | 4, 5 | 506 | 4911025 | ||
| CT010199.1 | - | 4, 5 | 431 | |||
| U83630.1 | - | 4, 5 | 520 | |||
| U83636.1 | - | 4, 5 | 431 | |||

Sp100
Genomic Map