siRNA Details
| Gene Symbol: Ccdc154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079929.2 | NP_001073398.2 | 13, 14 | 1867 | ||
| GenBank mRNA | AK133084.1 | - | 14, 15 | 1984 | ||
| BB017594.2 | - | 2, 3 | 128 | |||
| JN935900.1 | - | 12, 13 | 1486 | |||

Ccdc154
Genomic Map