siRNA Details
| Gene Symbol: Hectd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144788.2 | NP_659037.2 | 5 | 1182 | ||
| GenBank mRNA | AK042387.1 | - | 5 | 955 | ||
| AK153674.1 | - | 5 | 999 | |||
| AK169603.1 | - | 5 | 975 | |||
| AK172326.1 | - | 5 | 1183 | |||
| BC031446.1 | - | 5 | 922 | |||
| BC048868.1 | - | 11 | ||||

Hectd1
Genomic Map