siRNA Details
| Gene Symbol: Sptlc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011479.3 | NP_035609.1 | 4 | 549 | ||
| GenBank mRNA | AK044917.1 | - | 4 | 550 | ||
| AK057644.1 | - | 4 | 585 | |||
| AK089019.1 | - | 4 | 543 | |||
| AK159346.1 | - | 4 | 550 | |||
| BC003227.1 | - | 4 | 582 | 3483158 | ||
| U27455.1 | - | 4 | 517 | |||
| X95642.1 | - | 4 | 520 | |||

Sptlc2
Genomic Map