siRNA Details
| Gene Symbol: Ubald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145359.2 | NP_663334.1 | 1 | 199 | ||
| GenBank mRNA | BC013706.1 | - | 1 | 145 | 3987738 | |
| BC056472.1 | - | 1 | 153 | |||
| BC064081.1 | - | 1 | 150 | 1364938 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubald1| Gene Symbol: Ubald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145359.2 | NP_663334.1 | 1 | 199 | ||
| GenBank mRNA | BC013706.1 | - | 1 | 145 | 3987738 | |
| BC056472.1 | - | 1 | 153 | |||
| BC064081.1 | - | 1 | 150 | 1364938 | ||