siRNA Details
| Gene Symbol: Srebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033218.1 | NP_150087.1 | 7 | 1335 | ||
| GenBank mRNA | AF289715.2 | - | 5 | 662 | ||
| AF374267.1 | - | 7 | 1188 | |||
| AK155857.1 | - | 8 | 1283 | |||
| AK172219.1 | - | 7 | 1342 | |||
| AK197179.1 | - | 208 | ||||
| AK203075.1 | - | 3 | 208 | |||
| AK203491.1 | - | 3 | 209 | |||

Srebf2
Genomic Map