siRNA Details
| Gene Symbol: Srebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033218.1 | NP_150087.1 | 11, 12 | 2308 | ||
| GenBank mRNA | AF374267.1 | - | 11, 12 | 2161 | ||
| AK155857.1 | - | 12, 13 | 2256 | |||
| AK172219.1 | - | 11, 12 | 2315 | |||
| BC005777.1 | - | 1, 2 | 50 | |||
| BQ963055.1 | - | 2, 3 | 363 | |||

Srebf2
Genomic Map