siRNA Details
| Gene Symbol: Klhl25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122780.1 | NP_001116252.1 | 2 | 728 | ||
| NM_029652.1 | NP_083928.1 | 2 | 739 | |||
| NM_182782.2 | NP_877583.1 | 2 | 739 | |||
| GenBank mRNA | AK143086.1 | - | 2 | 685 | ||
| BC011221.1 | - | 2 | 670 | |||
| BC022930.1 | - | 2 | 670 | |||
| BC027373.1 | - | 2 | 741 | 4948451 | ||

Klhl25
Genomic Map