siRNA Details
| Gene Symbol: Btla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037719.2 | NP_001032808.2 | 6 | 896 | ||
| NM_177584.3 | NP_808252.1 | 6 | 893 | |||
| GenBank mRNA | AK041334.1 | - | 6 | 895 | ||
| AY293285.1 | - | 6 | 880 | |||
| BC108963.1 | - | 6 | 895 | 40046744 | ||
| BC108964.1 | - | 6 | 898 | 40046745 | ||

Btla
Genomic Map