siRNA Details
| Gene Symbol: Map6d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198599.2 | NP_941001.2 | 1 | 129 | ||
| GenBank mRNA | AK163656.1 | - | 1 | 93 | ||
| BB653005.1 | - | 1 | 130 | |||
| BC116220.1 | - | 1 | 92 | 40091320 | ||
| BC116221.1 | - | 1 | 92 | 40091321 | ||

Map6d1
Genomic Map