siRNA Details
| Gene Symbol: Tmem132c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175432.3 | NP_780641.2 | 3, 4 | 1228 | ||
| GenBank mRNA | AK028289.1 | - | 3, 4 | 1247 | ||
| AK028541.1 | - | 3, 4 | 1229 | |||
| AK035053.1 | - | 1, 2 | 126 | |||
| AK157823.1 | - | 3, 4 | 1229 | |||

Tmem132c
Genomic Map