siRNA Details
| Gene Symbol: Tmem132c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175432.3 | NP_780641.2 | 9 | 2852 | ||
| GenBank mRNA | AK028289.1 | - | 9 | 2871 | ||
| AK028541.1 | - | 9 | 2853 | |||
| AK035053.1 | - | 7 | 1750 | |||
| AK157823.1 | - | 9 | 2853 | |||
| AK163356.1 | - | 5 | 1420 | |||

Tmem132c
Genomic Map