siRNA Details
| Gene Symbol: Trove2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013835.2 | NP_038863.1 | 2 | 477 | ||
| GenBank mRNA | AK076167.1 | - | 2 | 454 | ||
| AK167379.1 | - | 2 | 418 | |||
| AK167555.1 | - | 2 | 418 | |||
| AK167645.1 | - | 2 | 418 | |||
| BC040368.1 | - | 1 | 348 | |||
| BC051974.1 | - | 1 | 354 | 5697399 | ||
| BC052380.1 | - | 1 | 354 | 5697399 | ||
| L81154.1 | - | 1 | 325 | |||
| U66843.1 | - | 2 | 404 | |||

Trove2
Genomic Map