siRNA Details
| Gene Symbol: Zfp871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172458.3 | NP_766046.3 | 4 | 392 | ||
| GenBank mRNA | AI119099.1 | - | 4 | 392 | ||
| AK029041.1 | - | 4 | 380 | |||
| AK122579.1 | - | 4 | 355 | |||
| BC043671.1 | - | 4 | 507 | 5040597 | ||

Zfp871
Genomic Map