siRNA Details
| Gene Symbol: Ssr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166480.1 | NP_001159952.1 | 2 | 344 | ||
| NM_009279.3 | NP_033305.1 | 2 | 226 | |||
| GenBank mRNA | AK003044.1 | - | 2 | 206 | ||
| AK007921.1 | - | 2 | 182 | |||
| AK013723.1 | - | 2 | 188 | |||
| AK142600.1 | - | 1 | 1773 | |||
| AK160089.1 | - | 2 | 192 | |||
| BC008172.1 | - | 2 | 147 | 3481948 | ||
| BC012971.1 | - | 1 | 1727 | |||
| BC019419.1 | - | 1 | 1677 | |||
| BI989805.1 | - | 2 | 344 | |||
| BQ747160.1 | - | 2 | 129 | |||
| CJ144362.1 | - | 2 | 231 | |||
| X90582.1 | - | 2 | 307 | |||

Ssr4
Genomic Map