siRNA Details
| Gene Symbol: Znrf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011483.2 | NP_035613.2 | 1 | 771 | ||
| GenBank mRNA | AB016984.1 | - | 1 | 705 | ||
| AK005843.1 | - | 1 | 720 | |||
| BC140986.1 | - | 1 | 654 | 9055598 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znrf4| Gene Symbol: Znrf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011483.2 | NP_035613.2 | 1 | 771 | ||
| GenBank mRNA | AB016984.1 | - | 1 | 705 | ||
| AK005843.1 | - | 1 | 720 | |||
| BC140986.1 | - | 1 | 654 | 9055598 | ||