siRNA Details
| Gene Symbol: Tmem212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164437.1 | NP_001157909.1 | 3 | 489 | ||
| GenBank mRNA | AK053134.1 | - | 3 | 489 | ||
| BC119391.1 | - | 3 | 489 | 8734140 | ||
| BC119393.1 | - | 3 | 489 | 8734142 | ||
| BE980542.1 | - | 2 | 241 | |||

Tmem212
Genomic Map