siRNA Details
| Gene Symbol: Etl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081006.1 | NP_001074475.1 | 20 | 4622 | ||
| NM_001177631.1 | NP_001171102.1 | 18 | 4615 | |||
| GenBank mRNA | AK147510.1 | - | 18 | 4524 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Etl4| Gene Symbol: Etl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081006.1 | NP_001074475.1 | 20 | 4622 | ||
| NM_001177631.1 | NP_001171102.1 | 18 | 4615 | |||
| GenBank mRNA | AK147510.1 | - | 18 | 4524 | ||