siRNA Details
| Gene Symbol: Etl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081006.1 | NP_001074475.1 | 22 | 5826 | ||
| NM_001177630.2 | NP_001171101.2 | 18 | 4127 | |||
| NM_001177631.1 | NP_001171102.1 | 20 | 5819 | |||
| NM_029895.4 | NP_084171.2 | 19 | 4322 | |||
| NM_178059.5 | NP_835160.2 | 19 | 4355 | |||
| GenBank mRNA | AB093289.2 | - | 14 | 2996 | ||
| AB125594.2 | - | 19 | 4355 | |||
| AB125595.2 | - | 19 | 4322 | |||
| AK147510.1 | - | 20 | 5728 | |||
| BC026657.1 | - | 7 | 1284 | |||
| BC055803.1 | - | 2 | 810 | |||
| BC158051.1 | - | 18 | 3849 | 9007405 | ||
| BC172143.1 | - | 18 | 3860 | |||

Etl4
Genomic Map