siRNA Details
| Gene Symbol: Aqp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177587.2 | NP_808255.1 | 1, 2 | 677 | ||
| GenBank mRNA | AB084104.1 | - | 1, 2 | 740 | ||
| BC107213.1 | - | 1, 2 | 669 | 40051613 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aqp12| Gene Symbol: Aqp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177587.2 | NP_808255.1 | 1, 2 | 677 | ||
| GenBank mRNA | AB084104.1 | - | 1, 2 | 740 | ||
| BC107213.1 | - | 1, 2 | 669 | 40051613 | ||