siRNA Details
Gene Symbol: Fanci | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145946.2 | NP_666058.2 | 19 | 1922 | ||
GenBank mRNA | AK085572.1 | - | 17 | 1772 | ||
BC027836.1 | - | 17 | 1649 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fanci | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145946.2 | NP_666058.2 | 19 | 1922 | ||
GenBank mRNA | AK085572.1 | - | 17 | 1772 | ||
BC027836.1 | - | 17 | 1649 |