siRNA Details
| Gene Symbol: Fanci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145946.2 | NP_666058.2 | 9 | 784 | ||
| GenBank mRNA | AK085572.1 | - | 7 | 634 | ||
| BC025462.1 | - | 9 | 818 | |||
| BC027836.1 | - | 9 | 780 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fanci| Gene Symbol: Fanci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145946.2 | NP_666058.2 | 9 | 784 | ||
| GenBank mRNA | AK085572.1 | - | 7 | 634 | ||
| BC025462.1 | - | 9 | 818 | |||
| BC027836.1 | - | 9 | 780 | |||