siRNA Details
| Gene Symbol: Myo5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081322.1 | NP_001074791.1 | 19 | 2365 | ||
| GenBank mRNA | AK049367.1 | - | 19 | 2360 | ||
| AK142326.1 | - | 20 | 2379 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo5c| Gene Symbol: Myo5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081322.1 | NP_001074791.1 | 19 | 2365 | ||
| GenBank mRNA | AK049367.1 | - | 19 | 2360 | ||
| AK142326.1 | - | 20 | 2379 | |||